Assay Details
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113062.4 | XP_001113062.1 | 270 | 94 | ||
XM_002802268.3 | XP_002802314.1 | 349 | 94 | |||
XM_028837247.1 | XP_028693080.1 | 231 | 94 | |||
XM_028837253.1 | XP_028693086.1 | 222 | 94 | |||
GenBank mRNA | JU344999.1 | - | 231 | 94 | ||
JU478322.1 | - | 262 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



