Assay Details
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134416.2 | XP_014989902.1 | 594 | 133 | ||
XM_015134417.2 | XP_014989903.1 | 615 | 133 | |||
XM_015134418.2 | XP_014989904.1 | 611 | 133 | |||
XM_015134419.2 | XP_014989905.1 | 448 | 133 | |||
XM_015134420.2 | XP_014989906.1 | 692 | 133 | |||
XM_015134421.2 | XP_014989907.1 | 688 | 133 | |||
XM_015134422.2 | XP_014989908.1 | 340 | 133 | |||
XM_028845565.1 | XP_028701398.1 | 895 | 133 | |||
XM_028845566.1 | XP_028701399.1 | 891 | 133 | |||
XM_028845567.1 | XP_028701400.1 | 726 | 133 | |||
XM_028845568.1 | XP_028701401.1 | 907 | 133 | |||
XM_028845569.1 | XP_028701402.1 | 484 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
