Assay Details
Gene Symbol: MED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260740.1 | NP_001247669.1 | 257 | 127 | ||
GenBank mRNA | JU322544.1 | - | 275 | 127 | ||
JU473224.1 | - | 269 | 127 | |||
JV044415.1 | - | 257 | 127 | |||
JV637267.1 | - | 275 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



