Assay Details
Gene Symbol: PCDHB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090927.4 | XP_001090927.1 | 5176 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
