Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134729.2 | XP_014990215.1 | 323 | 133 | ||
| XM_015134730.2 | XP_014990216.1 | 335 | 133 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015134729.2, XM_015134730.2 transcripts.
Set Membership: |
|

GPR85
Genomic Map