Assay Details
| Gene Symbol: LRRC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257576.1 | NP_001244505.1 | 2326 | 117 | ||
| XM_028834399.1 | XP_028690232.1 | 2478 | 117 | |||
| GenBank mRNA | JU335482.1 | - | 2326 | 117 | ||
| JU476427.1 | - | 2382 | 117 | |||
| JV047669.1 | - | 2288 | 117 | |||
More Information
Set Membership: |
|

LRRC8A
Genomic Map