Assay Details
| Gene Symbol: MYORG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194089.3 | NP_001181018.2 | 174 | 120 | ||
| GenBank mRNA | JV667247.1 | - | 86 | 120 | ||
| JV719471.1 | - | 103 | 120 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194089.3 transcripts.
Set Membership: |
|

MYORG
Genomic Map