Assay Details
Gene Symbol: RAP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193832.3 | NP_001180761.1 | 480 | 85 | ||
GenBank mRNA | JU328312.1 | - | 310 | 85 | ||
JU335754.1 | - | 387 | 85 | |||
JU471128.1 | - | 634 | 85 | |||
JU476637.1 | - | 381 | 85 | |||
JU476638.1 | - | 381 | 85 | |||
JV047822.1 | - | 351 | 85 | |||
JV720329.1 | - | 425 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
