Assay Details
| Gene Symbol: RAP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194044.2 | NP_001180973.1 | 1068 | 137 | ||
| XM_015128176.2 | XP_014983662.1 | 903 | 137 | |||
| GenBank mRNA | JU320733.1 | - | 581 | 137 | ||
| JU471431.1 | - | 559 | 137 | |||
| JV045617.1 | - | 714 | 137 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAP2C
Genomic Map