Assay Details
| Gene Symbol: ALG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190890.2 | NP_001177819.2 | 1206 | 78 | ||
| GenBank mRNA | JU334580.1 | - | 1213 | 78 | ||
| JU334581.1 | - | 1213 | 78 | |||
| JU334582.1 | - | 1213 | 78 | |||
| JU477990.1 | - | 1213 | 78 | |||
| JU496957.1 | - | 1213 | 78 | |||
More Information
Set Membership: |
|

ALG11
Genomic Map