Assay Details
| Gene Symbol: DMRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117824.2 | XP_014973310.2 | 543 | 77 | ||
| XM_015117825.2 | XP_014973311.2 | 499 | 77 | |||
| GenBank mRNA | JV053672.1 | - | 292 | 77 | ||
| JV635996.1 | - | 397 | 77 | |||
More Information
Set Membership: |
|

DMRT1
Genomic Map