Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260710.1 | NP_001247639.1 | 146 | 126 | ||
| GenBank mRNA | JU336542.1 | - | 165 | 126 | ||
| JU473181.1 | - | 129 | 126 | |||
| JV048339.1 | - | 146 | 126 | |||
| JV629981.1 | - | 181 | 126 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260710.1 transcripts.
Set Membership: |
|

GFOD2
Genomic Map