Assay Details
| Gene Symbol: LRRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132715.2 | XP_014988201.1 | 175 | 136 | ||
| GenBank mRNA | JV629920.1 | - | 146 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015132715.2 transcripts.
Set Membership: |
|

LRRC3
Genomic Map