Assay Details
| Gene Symbol: TRIM56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107405.4 | XP_001107405.2 | 341 | 149 | ||
| XM_015134514.2 | XP_014990000.1 | 555 | 149 | |||
| XM_028845613.1 | XP_028701446.1 | 199 | 149 | |||
| XM_028845614.1 | XP_028701447.1 | 291 | 149 | |||
| GenBank mRNA | JU323960.1 | - | 227 | 149 | ||
| JU472569.1 | - | 158 | 149 | |||
| JV043499.1 | - | 233 | 149 | |||
| JV638700.1 | - | 243 | 149 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001107405.4, XM_015134514.2, XM_028845613.1, XM_028845614.1 transcripts.
Set Membership: |
|

TRIM56
Genomic Map