Assay Details
| Gene Symbol: GNGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266578.1 | NP_001253507.1 | 209 | 129 | ||
| XM_015119234.2 | XP_014974720.1 | 332 | 129 | |||
| GenBank mRNA | JU320942.1 | - | 175 | 129 | ||
| JU473358.1 | - | 209 | 129 | |||
| JU473359.1 | - | 209 | 129 | |||
More Information
Set Membership: |
|

GNGT2
Genomic Map