Assay Details
Gene Symbol: TMEM121B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102577.4 | XP_001102577.2 | 1192 | 56 | ||
GenBank mRNA | JV613518.1 | - | 67 | 56 | ||
JV712322.1 | - | 155 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
