Assay Details
Gene Symbol: SLITRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093456.4 | XP_001093456.2 | 566 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001093456.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
