Assay Details
| Gene Symbol: PYM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257874.1 | NP_001244803.1 | 131 | 149 | ||
| XM_028828912.1 | XP_028684745.1 | 372 | 149 | |||
| XM_028828913.1 | XP_028684746.1 | 282 | 149 | |||
| GenBank mRNA | JU336707.1 | - | 131 | 149 | ||
| JU471933.1 | - | 139 | 149 | |||
| JV048440.1 | - | 229 | 149 | |||
More Information
Set Membership: |
|

PYM1
Genomic Map