Assay Details
| Gene Symbol: RHOXF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128106.2 | XP_014983592.1 | 798 | 92 | ||
| XM_028842444.1 | XP_028698277.1 | 4486 | 92 | |||
| XM_028842445.1 | XP_028698278.1 | 2552 | 92 | |||
| XM_028842446.1 | XP_028698279.1 | 3789 | 92 | |||
| GenBank mRNA | JV636413.1 | - | 692 | 92 | ||
More Information
Set Membership: |
|

RHOXF2
Genomic Map