Assay Details
Gene Symbol: IGSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260991.1 | NP_001247920.1 | 158 | 116 | ||
XM_015125791.2 | XP_014981277.2 | 491 | 116 | |||
GenBank mRNA | JU336804.1 | - | 158 | 116 | ||
JV596529.1 | - | 481 | 116 | |||
JV631555.1 | - | 483 | 116 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
