Assay Details
Gene Symbol: LSM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257958.1 | NP_001244887.1 | 88 | 112 | ||
GenBank mRNA | JU336805.1 | - | 88 | 112 | ||
JU477559.1 | - | 132 | 112 | |||
JV560957.1 | - | 90 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257958.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
