Assay Details
Gene Symbol: GAL3ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111547.3 | XP_001111547.2 | 265 | 152 | ||
GenBank mRNA | JV600327.1 | - | 177 | 152 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001111547.3 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
