Assay Details
Gene Symbol: GAL3ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111547.3 | XP_001111547.2 | 505 | 140 | ||
GenBank mRNA | JV600327.1 | - | 417 | 140 | ||
JV700371.1 | - | 312 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
