Assay Details
Gene Symbol: GK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193898.1 | NP_001180827.1 | 128 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193898.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
