Assay Details
Gene Symbol: LRRC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093813.4 | XP_001093813.1 | 597 | 140 | ||
GenBank mRNA | JU320950.1 | - | 253 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001093813.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
