Assay Details
| Gene Symbol: DEFB118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037344.1 | NP_001032421.1 | 81 | 104 | ||
| GenBank mRNA | AF207834.1 | - | 81 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001037344.1 transcripts.
Set Membership: |
|

DEFB118
Genomic Map