Assay Details
| Gene Symbol: CDKN2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266314.1 | NP_001253243.1 | 245 | 114 | ||
| XM_015139180.2 | XP_014994666.1 | 2127 | 114 | |||
| GenBank mRNA | JU332475.1 | - | 453 | 114 | ||
| JU332476.1 | - | 205 | 114 | |||
| JU471305.1 | - | 1325 | 114 | |||
| JU473943.1 | - | 245 | 114 | |||
More Information
Set Membership: |
|

CDKN2C
Genomic Map