Assay Details
| Gene Symbol: SLC32A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266377.1 | NP_001253306.1 | 599 | 108 | ||
| GenBank mRNA | JU321903.1 | - | 599 | 108 | ||
| JV528639.1 | - | 621 | 108 | |||
| JV565759.1 | - | 648 | 108 | |||
More Information
Set Membership: |
|

SLC32A1
Genomic Map