Assay Details
Gene Symbol: LOC713067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128980.2 | XP_014984466.2 | 322 | 125 | ||
GenBank mRNA | JV046194.1 | - | 222 | 125 | ||
JV046195.1 | - | 222 | 125 | |||
JV635956.1 | - | 320 | 125 | |||
JV638324.1 | - | 320 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015128980.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
