Assay Details
Gene Symbol: SPSB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132000.2 | XP_014987486.1 | 764 | 81 | ||
XM_015132001.2 | XP_014987487.1 | 600 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015132000.2, XM_015132001.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
