Assay Details
Gene Symbol: MARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086799.4 | XP_001086799.3 | 129 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
