Assay Details
| Gene Symbol: NSMCE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257507.1 | NP_001244436.1 | 53 | 104 | ||
| GenBank mRNA | JU335479.1 | - | 53 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257507.1 transcripts.
Set Membership: |
|

NSMCE3
Genomic Map