Assay Details
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141642.2 | XP_014997128.1 | 382 | 102 | ||
XM_015141643.2 | XP_014997129.1 | 405 | 102 | |||
XM_015141646.2 | XP_014997132.1 | 382 | 102 | |||
XM_015141647.2 | XP_014997133.1 | 382 | 102 | |||
XM_015141648.2 | XP_014997134.1 | 367 | 102 | |||
XM_028850019.1 | XP_028705852.1 | 301 | 102 | |||
GenBank mRNA | JU319957.1 | - | 288 | 102 | ||
JU319958.1 | - | 288 | 102 | |||
JU319959.1 | - | 288 | 102 | |||
JU476415.1 | - | 269 | 102 | |||
JV047658.1 | - | 288 | 102 | |||
JV047659.1 | - | 288 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
