Assay Details
Gene Symbol: HEXIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194678.1 | NP_001181607.1 | 278 | 145 | ||
XM_015119771.2 | XP_014975257.1 | 2462 | 145 | |||
XM_015119772.2 | XP_014975258.1 | 2459 | 145 | |||
XM_015119773.2 | XP_014975259.1 | 2964 | 145 | |||
XM_015119777.2 | XP_014975263.1 | 2767 | 145 | |||
XM_015119778.1 | XP_014975264.1 | 711 | 145 | |||
XM_028835707.1 | XP_028691540.1 | 2967 | 145 | |||
XM_028835708.1 | XP_028691541.1 | 2872 | 145 | |||
XM_028835709.1 | XP_028691542.1 | 2875 | 145 | |||
XM_028835710.1 | XP_028691543.1 | 2954 | 145 | |||
XM_028835711.1 | XP_028691544.1 | 2862 | 145 | |||
XM_028835712.1 | XP_028691545.1 | 2449 | 145 | |||
GenBank mRNA | JU322753.1 | - | 125 | 145 | ||
JV706707.1 | - | 126 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
