Assay Details
Gene Symbol: CCDC127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194083.3 | NP_001181012.2 | 189 | 133 | ||
GenBank mRNA | JU332964.1 | - | 206 | 133 | ||
JU474339.1 | - | 196 | 133 | |||
JV046216.1 | - | 193 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
