Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092987.3 | XP_001092987.3 | 1017 | 137 | ||
XM_001093102.4 | XP_001093102.3 | 1013 | 137 | |||
XM_015149644.2 | XP_015005130.2 | 904 | 137 | |||
XM_028847320.1 | XP_028703153.1 | 1995 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
