Assay Details
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100403.4 | XP_001100403.1 | 579 | 141 | ||
XM_015138960.2 | XP_014994446.1 | 579 | 141 | |||
XM_015138964.2 | XP_014994450.1 | 579 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001100403.4, XM_015138960.2, XM_015138964.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
