Assay Details
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100403.4 | XP_001100403.1 | 757 | 123 | ||
XM_028848090.1 | XP_028703923.1 | 182 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001100403.4, XM_028848090.1 transcripts.
Set Membership: |
![]() ![]() |
