Assay Details
Gene Symbol: PWWP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127956.2 | XP_014983442.1 | 878 | 69 | ||
GenBank mRNA | JV046230.1 | - | 101 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015127956.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
