Assay Details
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193740.2 | NP_001180669.1 | 1443 | 89 | ||
XM_002804165.3 | XP_002804211.1 | 1524 | 89 | |||
XM_002804166.3 | XP_002804212.1 | 1279 | 89 | |||
XM_015138948.2 | XP_014994434.1 | 1545 | 89 | |||
XM_015138949.2 | XP_014994435.1 | 1513 | 89 | |||
XM_015138954.2 | XP_014994440.1 | 1604 | 89 | |||
XM_028847974.1 | XP_028703807.1 | 1516 | 89 | |||
XM_028847975.1 | XP_028703808.1 | 1556 | 89 | |||
XM_028847976.1 | XP_028703809.1 | 1553 | 89 | |||
XM_028847977.1 | XP_028703810.1 | 1664 | 89 | |||
XM_028847978.1 | XP_028703811.1 | 1446 | 89 | |||
GenBank mRNA | JU474523.1 | - | 1327 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
