Assay Details
| Gene Symbol: CLEC14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091540.4 | XP_001091540.3 | 213 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001091540.4 transcripts.
Set Membership: |
|

CLEC14A
Genomic Map