Assay Details
| Gene Symbol: FAM181B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194034.2 | NP_001180963.1 | 1681 | 86 | ||
| GenBank mRNA | JU333387.1 | - | 1628 | 86 | ||
| JV460223.1 | - | 671 | 86 | |||
More Information
Set Membership: |
|

FAM181B
Genomic Map