Assay Details
| Gene Symbol: KCTD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125960.2 | XP_014981446.1 | 454 | 91 | ||
| XM_028841521.1 | XP_028697354.1 | 454 | 91 | |||
| GenBank mRNA | JU322113.1 | - | 427 | 91 | ||
| JU473550.1 | - | 413 | 91 | |||
| JV043692.1 | - | 424 | 91 | |||
| JV529986.1 | - | 394 | 91 | |||
| JV577192.1 | - | 432 | 91 | |||
More Information
Set Membership: |
|

KCTD13
Genomic Map