Assay Details
| Gene Symbol: PKIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193423.2 | NP_001180352.2 | 171 | 134 | ||
| XM_015137324.2 | XP_014992810.2 | 696 | 134 | |||
| XM_015137326.2 | XP_014992812.2 | 260 | 134 | |||
| XM_015137327.2 | XP_014992813.2 | 798 | 134 | |||
| XM_015137328.2 | XP_014992814.2 | 733 | 134 | |||
| XM_015137329.2 | XP_014992815.2 | 601 | 134 | |||
| XM_028846846.1 | XP_028702679.1 | 749 | 134 | |||
| XM_028846847.1 | XP_028702680.1 | 359 | 134 | |||
| XM_028846848.1 | XP_028702681.1 | 390 | 134 | |||
| GenBank mRNA | JU335396.1 | - | 318 | 134 | ||
| JV586897.1 | - | 211 | 134 | |||
More Information
Set Membership: |
|

PKIB
Genomic Map