Assay Details
Gene Symbol: LOC114674356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840866.1 | XP_028696699.1 | 658 | 135 | ||
XM_028840867.1 | XP_028696700.1 | 325 | 135 | |||
XM_028840868.1 | XP_028696701.1 | 306 | 135 |
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV645451.1 | - | 207 | 135 |

More Information
Set Membership: |
![]() ![]() |
