Assay Details
| Gene Symbol: APOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834006.1 | XP_028689839.1 | 429 | 150 | ||
| GenBank mRNA | JV059699.1 | - | 144 | 150 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

APOA1
Genomic Map