Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195417.1 | NP_001182346.1 | 457 | 101 | ||
GenBank mRNA | AY369789.1 | - | 103 | 101 | ||
CO726823.1 | - | 501 | 101 | |||
JU336288.1 | - | 588 | 101 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
