Assay Details
Gene Symbol: CSTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257698.1 | NP_001244627.1 | 158 | 160 | ||
GenBank mRNA | JU323350.1 | - | 158 | 160 | ||
JU474405.1 | - | 114 | 160 | |||
JV045152.1 | - | 158 | 160 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257698.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
