Assay Details
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128374.2 | XP_014983860.2 | 1390 | 137 | ||
XM_015128375.2 | XP_014983861.2 | 1399 | 137 | |||
XM_015128376.2 | XP_014983862.2 | 1347 | 137 | |||
XM_028842053.1 | XP_028697886.1 | 1202 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
