Assay Details
Gene Symbol: POU1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042860.1 | NP_001036325.1 | 325 | 107 | ||
XM_015128786.2 | XP_014984272.1 | 107 | ||||
GenBank mRNA | U53566.1 | - | 325 | 107 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
